siRNA Details
| Gene Symbol: SMIM15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001048249.3 | NP_001041714.1 | 3 | 1094 | ||
| GenBank mRNA | BC014242.2 | - | 3 | 948 | ||
| BC047489.2 | - | 3 | 975 | |||
| BC066550.1 | - | 3 | 1073 | |||
| BC103711.1 | - | 3 | 1049 | |||
| BX538015.1 | - | 3 | 1089 | |||
| BX648459.1 | - | 3 | 1066 | |||

SMIM15
Genomic Map