siRNA Details
| Gene Symbol: EHMT1-IT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024341.1 | 1 | 1605 | |||
| GenBank mRNA | AK097611.1 | - | 1 | 1614 | ||
| AK127990.1 | - | 1 | 1614 | |||
| BX106199.1 | - | 1 | 387 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

EHMT1-IT1| Gene Symbol: EHMT1-IT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024341.1 | 1 | 1605 | |||
| GenBank mRNA | AK097611.1 | - | 1 | 1614 | ||
| AK127990.1 | - | 1 | 1614 | |||
| BX106199.1 | - | 1 | 387 | |||