siRNA Details
| Gene Symbol: CT47B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145718.1 | NP_001139190.1 | 1 | 674 | ||
| XM_017029734.1 | XP_016885223.1 | 1 | 636 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CT47B1| Gene Symbol: CT47B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145718.1 | NP_001139190.1 | 1 | 674 | ||
| XM_017029734.1 | XP_016885223.1 | 1 | 636 | |||