siRNA Details
| Gene Symbol: LOC643441 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038846.1 | 2, 3 | 273 | |||
| GenBank mRNA | AK123703.1 | - | 2, 3 | 273 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC643441| Gene Symbol: LOC643441 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038846.1 | 2, 3 | 273 | |||
| GenBank mRNA | AK123703.1 | - | 2, 3 | 273 | ||