siRNA Details
| Gene Symbol: BRDTP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003539.1 | 1 | 366 | |||
| GenBank mRNA | AK124282.1 | - | 1 | 2061 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BRDTP1| Gene Symbol: BRDTP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003539.1 | 1 | 366 | |||
| GenBank mRNA | AK124282.1 | - | 1 | 2061 | ||