siRNA Details
Gene Symbol: BRDT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242805.2 | NP_001229734.2 | 18 | 2904 | ||
NM_001242806.2 | NP_001229735.2 | 17 | 2847 | |||
NM_001242807.2 | NP_001229736.2 | 16 | 2503 | |||
NM_001242808.2 | NP_001229737.2 | 16 | 2697 | |||
NM_001242810.2 | NP_001229739.2 | 17 | 3191 | |||
NM_001726.4 | NP_001717.3 | 17 | 2630 | |||
NM_207189.3 | NP_997072.2 | 17 | 2835 | |||
XM_006710853.3 | XP_006710916.1 | 18 | 3420 | |||
XM_006710854.3 | XP_006710917.1 | 19 | 3265 | |||
XM_006710855.3 | XP_006710918.1 | 19 | 3270 | |||
XM_006710856.3 | XP_006710919.1 | 19 | 3244 | |||
XM_006710857.3 | XP_006710920.1 | 19 | 3234 | |||
XM_011542032.2 | XP_011540334.1 | 19 | 2754 | |||
XM_011542033.2 | XP_011540335.1 | 19 | 2728 | |||
XM_011542034.2 | XP_011540336.1 | 19 | 2744 | |||
XM_011542035.2 | XP_011540337.1 | 19 | 2718 | |||
XM_011542036.2 | XP_011540338.1 | 17 | 2633 | |||
GenBank mRNA | AF019085.1 | - | 17 | 2593 | ||
AK302758.1 | - | 16 | 2439 | |||
AK303008.1 | - | 17 | 2595 | |||
AK316442.1 | - | 16 | 2502 | |||
AY338951.1 | - | 18 | 2904 | |||
BX648135.1 | - | 17 | 2926 |
Gene Symbol: BRDTP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003539.1 | 1 | 315 | |||
GenBank mRNA | AK124282.1 | - | 1 | 2010 |
