siRNA Details
| Gene Symbol: MAP4K3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270425.1 | NP_001257354.1 | 1, 2 | 429 | ||
| NM_003618.3 | NP_003609.2 | 1, 2 | 429 | |||
| GenBank mRNA | AK291467.1 | - | 1, 2 | 395 | ||
| AK314487.1 | - | 1, 2 | 266 | |||
| BC071579.1 | - | 1, 2 | 421 | 5296205 | ||
| DA409370.1 | - | 1, 2 | 431 | |||

MAP4K3
Genomic Map