siRNA Details
| Gene Symbol: ERICH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290030.1 | NP_001276959.1 | 3, 4 | 585 | ||
| NM_001290031.1 | NP_001276960.1 | 3, 4 | 585 | |||
| NR_110644.1 | 4, 5 | 892 | ||||
| XM_011510998.2 | XP_011509300.1 | 5, 6 | 717 | |||
| XM_011510999.2 | XP_011509301.1 | 4, 5 | 384 | |||
| XM_011511001.2 | XP_011509303.1 | 3, 4 | 1063 | |||
| XM_017003875.1 | XP_016859364.1 | 4, 5 | 1086 | |||
| XM_017003876.1 | XP_016859365.1 | 3, 4 | 101 | |||
| XM_017003877.1 | XP_016859366.1 | 4, 5 | 384 | |||
| GenBank mRNA | AA401652.1 | - | 3, 4 | 136 | ||
| BQ269226.1 | - | 4, 5 | 461 | |||

ERICH2
Genomic Map