siRNA Details
| Gene Symbol: LOC643770 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038383.1 | 3, 4 | 172 | |||
| GenBank mRNA | AK126817.1 | - | 3, 4 | 172 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC643770| Gene Symbol: LOC643770 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038383.1 | 3, 4 | 172 | |||
| GenBank mRNA | AK126817.1 | - | 3, 4 | 172 | ||