siRNA Details
| Gene Symbol: KRTAP24-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085455.2 | NP_001078924.1 | 1 | 472 | ||
| GenBank mRNA | AB096935.1 | - | 1 | 472 | ||
| AM238513.1 | - | 1 | 446 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRTAP24-1| Gene Symbol: KRTAP24-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085455.2 | NP_001078924.1 | 1 | 472 | ||
| GenBank mRNA | AB096935.1 | - | 1 | 472 | ||
| AM238513.1 | - | 1 | 446 | |||