siRNA Details
Gene Symbol: CBLN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039771.2 | NP_001034860.1 | 2 | 850 | ||
GenBank mRNA | AY359070.1 | - | 2 | 850 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CBLN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039771.2 | NP_001034860.1 | 2 | 850 | ||
GenBank mRNA | AY359070.1 | - | 2 | 850 |