siRNA Details
| Gene Symbol: ZNF733P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003952.1 | 3 | 730 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF733P| Gene Symbol: ZNF733P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003952.1 | 3 | 730 | |||