siRNA Details
| Gene Symbol: CBWD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085457.1 | NP_001078926.1 | 8 | 757 | ||
| XM_017015019.1 | XP_016870508.1 | 7 | 757 | |||
| XM_017015020.1 | XP_016870509.1 | 7 | 757 | |||
| XM_017015021.1 | XP_016870510.1 | 7 | 757 | |||
| XM_017015023.1 | XP_016870512.1 | 7 | 757 | |||
| XM_017015026.1 | XP_016870515.1 | 7 | 757 | |||
| XM_017015027.1 | XP_016870516.1 | 7 | 757 | |||
| XM_017015028.1 | XP_016870517.1 | 8 | 625 | |||
| XM_017015029.1 | XP_016870518.1 | 4 | 576 | |||
| XM_017015030.1 | XP_016870519.1 | 7 | 757 | |||
| XM_017015031.1 | XP_016870520.1 | 3 | 268 | |||
| XM_017015032.1 | XP_016870521.1 | 7 | 757 | |||
| XM_017015034.1 | XP_016870523.1 | 2 | 131 | |||
| XM_017015035.1 | XP_016870524.1 | 8 | 817 | |||
| GenBank mRNA | AF293368.1 | - | 1 | 62 | ||

CBWD6
Genomic Map