siRNA Details
| Gene Symbol: CBWD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085457.1 | NP_001078926.1 | 12 | 946 | ||
| XM_017015019.1 | XP_016870508.1 | 12 | 991 | |||
| XM_017015020.1 | XP_016870509.1 | 11 | 946 | |||
| XM_017015021.1 | XP_016870510.1 | 11 | 934 | |||
| XM_017015022.1 | XP_016870511.1 | 11 | 904 | |||
| XM_017015023.1 | XP_016870512.1 | 10 | 889 | |||
| XM_017015024.1 | XP_016870513.1 | 10 | 859 | |||
| XM_017015025.1 | XP_016870514.1 | 10 | 847 | |||
| XM_017015026.1 | XP_016870515.1 | 12 | 991 | |||
| XM_017015028.1 | XP_016870517.1 | 13 | 859 | |||
| XM_017015029.1 | XP_016870518.1 | 9 | 810 | |||
| XM_017015031.1 | XP_016870520.1 | 8 | 502 | |||
| XM_017015033.1 | XP_016870522.1 | 7 | 415 | |||
| XM_017015034.1 | XP_016870523.1 | 7 | 365 | |||
| XM_017015035.1 | XP_016870524.1 | 11 | 949 | |||
| XM_017015036.1 | XP_016870525.1 | 10 | 862 | |||
| GenBank mRNA | AF293368.1 | - | 5 | 251 | ||
| AK293173.1 | - | 12 | 995 | |||
| BC069006.1 | - | 13 | 1087 | |||
| CR627025.1 | - | 1 | 1130 | |||

CBWD6
Genomic Map