siRNA Details
| Gene Symbol: CTSLP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033407.1 | 4 | 448 | |||
| GenBank mRNA | AK309109.1 | - | 4 | 448 | ||
| Gene Symbol: CTSLP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033405.1 | 1 | 86 | |||
| GenBank mRNA | DA786334.1 | - | 1 | 87 | ||

CTSLP2
Genomic Map