siRNA Details
| Gene Symbol: IRX4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278632.1 | NP_001265561.1 | 6 | 2067 | ||
| NM_001278633.1 | NP_001265562.1 | 7 | 2145 | |||
| NM_001278634.1 | NP_001265563.1 | 6 | 2072 | |||
| NM_001278635.1 | NP_001265564.1 | 7 | 2150 | |||
| NM_016358.2 | NP_057442.1 | 5 | 1968 | |||
| GenBank mRNA | AB690778.1 | - | 6 | 2067 | ||
| AB690779.1 | - | 7 | 2145 | |||
| AB690780.1 | - | 6 | 2072 | |||
| AB690781.1 | - | 7 | 2150 | |||
| BC110912.1 | - | 5 | 1856 | 3450161 | ||

IRX4
Genomic Map