siRNA Details
| Gene Symbol: SDHAF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042631.2 | NP_001036096.1 | 1 | 1095 | ||
| GenBank mRNA | BC007205.1 | - | 1 | 1084 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SDHAF1| Gene Symbol: SDHAF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042631.2 | NP_001036096.1 | 1 | 1095 | ||
| GenBank mRNA | BC007205.1 | - | 1 | 1084 | ||