siRNA Details
| Gene Symbol: ARL14EPL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195581.1 | NP_001182510.1 | 3 | 331 | ||
| XM_017009722.1 | XP_016865211.1 | 4 | 425 | |||
| GenBank mRNA | DN930361.1 | - | 2 | 121 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ARL14EPL| Gene Symbol: ARL14EPL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195581.1 | NP_001182510.1 | 3 | 331 | ||
| XM_017009722.1 | XP_016865211.1 | 4 | 425 | |||
| GenBank mRNA | DN930361.1 | - | 2 | 121 | ||