siRNA Details
Gene Symbol: SYCE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001123225.1 | NP_001116697.1 | 2 | 85 | ||
GenBank mRNA | BC127859.1 | - | 1 | 70 | 40134492 | |
BC127860.1 | - | 1 | 70 | 40134499 | ||
EU082028.1 | - | 1 | 43 |
