siRNA Details
| Gene Symbol: LOC644285 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_130929.1 | 1 | 4149 | |||
| GenBank mRNA | AK054805.1 | - | 1 | 1290 | ||
| AK055011.1 | - | 1 | 542 | |||
| AK126853.1 | - | 1 | 4150 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC644285| Gene Symbol: LOC644285 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_130929.1 | 1 | 4149 | |||
| GenBank mRNA | AK054805.1 | - | 1 | 1290 | ||
| AK055011.1 | - | 1 | 542 | |||
| AK126853.1 | - | 1 | 4150 | |||