siRNA Details
| Gene Symbol: FMN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277313.1 | NP_001264242.1 | 4 | 713 | ||
| NM_001277314.1 | NP_001264243.1 | 4 | 713 | |||
| XM_011521504.2 | XP_011519806.1 | 4 | 536 | |||
| XM_011521505.2 | XP_011519807.1 | 4 | 745 | |||
| XM_011521506.2 | XP_011519808.1 | 4 | 744 | |||
| XM_011521507.2 | XP_011519809.1 | 4 | 745 | |||
| XM_017022130.1 | XP_016877619.1 | 1 | 907 | |||
| XM_017022131.1 | XP_016877620.1 | 4 | 745 | |||
| XM_017022133.1 | XP_016877622.1 | 4 | 742 | |||
| XM_017022135.1 | XP_016877624.1 | 4 | 742 | |||
| GenBank mRNA | CR749487.1 | - | 4 | 677 | ||

FMN1
Genomic Map