siRNA Details
| Gene Symbol: FMN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277314.1 | NP_001264243.1 | 5 | 2434 | ||
| GenBank mRNA | CR749487.1 | - | 5 | 2398 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FMN1| Gene Symbol: FMN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277314.1 | NP_001264243.1 | 5 | 2434 | ||
| GenBank mRNA | CR749487.1 | - | 5 | 2398 | ||