siRNA Details
| Gene Symbol: KRTAP2-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033184.3 | NP_149440.1 | 1 | 629 | ||
| GenBank mRNA | AJ406930.1 | - | 1 | 629 | ||
| BC101147.2 | - | 1 | 625 | 40015180 | ||
| BC101148.2 | - | 1 | 625 | 40015187 | ||
| BC101149.2 | - | 1 | 625 | 40015188 | ||

KRTAP2-4
Genomic Map