siRNA Details
| Gene Symbol: RAD21-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033886.1 | 1 | 414 | |||
| GenBank mRNA | BC031280.1 | - | 1 | 414 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RAD21-AS1| Gene Symbol: RAD21-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033886.1 | 1 | 414 | |||
| GenBank mRNA | BC031280.1 | - | 1 | 414 | ||