siRNA Details
| Gene Symbol: CDRT15L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190790.1 | NP_001177719.1 | 2 | 924 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CDRT15L2| Gene Symbol: CDRT15L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190790.1 | NP_001177719.1 | 2 | 924 | ||