siRNA Details
| Gene Symbol: FAM83G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039999.2 | NP_001035088.2 | 5 | 1921 | ||
| XM_017024953.1 | XP_016880442.1 | 5 | 2005 | |||
| GenBank mRNA | AK123558.1 | - | 3 | 2191 | ||
| BX402942.2 | - | 1 | 635 | |||

FAM83G
Genomic Map