siRNA Details
| Gene Symbol: FAM83G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039999.2 | NP_001035088.2 | 6 | 2517 | ||
| XM_017024953.1 | XP_016880442.1 | 6 | 2601 | |||
| GenBank mRNA | BC105988.1 | - | 2 | 501 | ||
| CR735308.1 | - | 1 | 76 | |||
| CX165458.1 | - | 2 | 304 | |||

FAM83G
Genomic Map