siRNA Details
| Gene Symbol: MEIG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080836.2 | NP_001074305.1 | 2 | 255 | ||
| GenBank mRNA | BG189536.1 | - | 1 | 139 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MEIG1| Gene Symbol: MEIG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080836.2 | NP_001074305.1 | 2 | 255 | ||
| GenBank mRNA | BG189536.1 | - | 1 | 139 | ||