siRNA Details
| Gene Symbol: GOLGA8M | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282468.1 | NP_001269397.1 | 19 | 2598 | ||
| XM_005268284.4 | XP_005268341.1 | 19 | 3397 | |||
| XM_005268285.4 | XP_005268342.1 | 18 | 3328 | |||
| XM_006720654.3 | XP_006720717.1 | 19 | 2541 | |||
| XM_011521951.2 | XP_011520253.1 | 20 | 2966 | |||
| XM_011521954.2 | XP_011520256.1 | 19 | 2531 | |||
| Gene Symbol: GOLGA8S | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038843.1 | 18 | 2865 | |||

GOLGA8M
Genomic Map