siRNA Details
| Gene Symbol: LOC107983967 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017003062.1 | XP_016858551.1 | 5, 6 | 572 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC107983967| Gene Symbol: LOC107983967 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017003062.1 | XP_016858551.1 | 5, 6 | 572 | ||