siRNA Details
| Gene Symbol: LINC00693 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038840.1 | 4 | 581 | |||
| GenBank mRNA | AA434563.1 | - | 2 | 117 | ||
| BX333929.2 | - | 2 | 261 | |||
| R20280.1 | - | 1 | 44 | |||
| R20979.1 | - | 1 | 57 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00693| Gene Symbol: LINC00693 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038840.1 | 4 | 581 | |||
| GenBank mRNA | AA434563.1 | - | 2 | 117 | ||
| BX333929.2 | - | 2 | 261 | |||
| R20280.1 | - | 1 | 44 | |||
| R20979.1 | - | 1 | 57 | |||