siRNA Details
| Gene Symbol: EIF3J-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034170.1 | 1 | 702 | |||
| NR_034171.1 | 2 | 237 | ||||
| GenBank mRNA | AK092547.1 | - | 1 | 453 | ||
| BC026043.1 | - | 1 | 690 | |||
| BC042851.1 | - | 1 | 456 | |||
| BC066970.1 | - | 1 | 702 | |||
| DB480720.1 | - | 2 | 237 | |||

EIF3J-AS1
Genomic Map