siRNA Details
| Gene Symbol: LRCOL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195520.1 | NP_001182449.1 | 6 | 803 | ||
| XM_011534780.2 | XP_011533082.1 | 5 | 1652 | |||
| XM_011534784.2 | XP_011533086.1 | 4 | 1510 | |||
| XM_011534785.2 | XP_011533087.1 | 4 | 1515 | |||
| XM_017018659.1 | XP_016874148.1 | 4 | 1628 | |||
| XM_017018660.1 | XP_016874149.1 | 5 | 779 | |||
| XM_017018661.1 | XP_016874150.1 | 4 | 1870 | |||
| XM_017018662.1 | XP_016874151.1 | 4 | 1613 | |||
| GenBank mRNA | AV653338.1 | - | 3 | 379 | ||
| JQ012741.1 | - | 6 | 726 | |||
| JQ012742.1 | - | 6 | 858 | |||

LRCOL1
Genomic Map