siRNA Details
| Gene Symbol: LINC00461 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015436.1 | 5 | 540 | |||
| GenBank mRNA | DA122372.1 | - | 5 | 456 | ||
| DA778665.1 | - | 5 | 544 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00461| Gene Symbol: LINC00461 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015436.1 | 5 | 540 | |||
| GenBank mRNA | DA122372.1 | - | 5 | 456 | ||
| DA778665.1 | - | 5 | 544 | |||