siRNA Details
| Gene Symbol: LOC101930094 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135518.1 | 3 | 368 | |||
| GenBank mRNA | AL551431.3 | - | 2 | 760 | ||
| BX111242.1 | - | 3 | 368 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101930094| Gene Symbol: LOC101930094 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135518.1 | 3 | 368 | |||
| GenBank mRNA | AL551431.3 | - | 2 | 760 | ||
| BX111242.1 | - | 3 | 368 | |||