siRNA Details
| Gene Symbol: CCDC179 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195637.1 | NP_001182566.1 | 3 | 131 | ||
| XM_011519802.2 | XP_011518104.1 | 3 | 676 | |||
| XM_011519803.2 | XP_011518105.1 | 3 | 664 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCDC179| Gene Symbol: CCDC179 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195637.1 | NP_001182566.1 | 3 | 131 | ||
| XM_011519802.2 | XP_011518104.1 | 3 | 676 | |||
| XM_011519803.2 | XP_011518105.1 | 3 | 664 | |||