siRNA Details
Gene Symbol: LOC102723778 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125935.1 | 5 | 462 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC102723778 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125935.1 | 5 | 462 |