siRNA Details
| Gene Symbol: GBAS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001202469.1 | NP_001189398.1 | 4 | 377 | ||
| NM_001483.2 | NP_001474.1 | 6 | 494 | |||
| GenBank mRNA | AF029786.1 | - | 6 | 469 | ||
| AJ001259.1 | - | 6 | 461 | |||
| AK097049.1 | - | 4 | 363 | |||
| AK125036.1 | - | 3 | 2750 | |||
| AK315648.1 | - | 6 | 479 | |||
| BC000732.2 | - | 6 | 467 | 2959662 | ||
| BC001837.2 | - | 6 | 467 | 2959662 | ||
| BC030821.1 | - | 6 | 476 | 4808962 | ||
| BT007112.1 | - | 6 | 461 | |||
| CR407612.1 | - | 6 | 461 | |||
| CX166452.1 | - | 6 | 494 | |||

GBAS
Genomic Map