siRNA Details
| Gene Symbol: LOC645752 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027024.1 | 9, 10 | 787 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC645752| Gene Symbol: LOC645752 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027024.1 | 9, 10 | 787 | |||