siRNA Details
| Gene Symbol: USP17L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256873.1 | NP_001243802.1 | 1 | 1390 | ||
| Gene Symbol: USP17L3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256871.1 | NP_001243800.1 | 1 | 1390 | ||

USP17L1
Genomic Map