siRNA Details
| Gene Symbol: TMEM14EP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_132656.1 | 1 | 352 | |||
| GenBank mRNA | AW866456.1 | - | 1 | 79 | ||
| AY358260.1 | - | 1 | 352 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM14EP| Gene Symbol: TMEM14EP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_132656.1 | 1 | 352 | |||
| GenBank mRNA | AW866456.1 | - | 1 | 79 | ||
| AY358260.1 | - | 1 | 352 | |||