siRNA Details
| Gene Symbol: NHSL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013627.2 | NP_001013649.2 | 8 | 3426 | ||
| XM_011530934.2 | XP_011529236.1 | 8 | 8218 | |||
| XM_017029473.1 | XP_016884962.1 | 8 | 7188 | |||
| XM_017029474.1 | XP_016884963.1 | 8 | 3330 | |||
| XM_017029475.1 | XP_016884964.1 | 4 | 6573 | |||
| XM_017029476.1 | XP_016884965.1 | 4 | 2690 | |||
| XM_017029477.1 | XP_016884966.1 | 4 | 7604 | |||

NHSL2
Genomic Map