siRNA Details
| Gene Symbol: RPS28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK293636.1 | - | 3 | 549 | ||
| AU126783.1 | - | 4 | 300 | |||
| BC000354.1 | - | 1 | 286 | 2964364 | ||
| BC018810.1 | - | 1 | 286 | |||
| BC070217.1 | - | 1 | 300 | 4248660 | ||
| BC070218.1 | - | 1 | 297 | 6649143 | ||
| L05091.1 | - | 4 | 284 | |||
| U58682.1 | - | 4 | 281 | |||

RPS28
Genomic Map