siRNA Details
| Gene Symbol: LINC00607 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037195.1 | 4 | 546 | |||
| GenBank mRNA | AK091865.1 | - | 4 | 445 | ||
| AK124897.1 | - | 4 | 546 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00607| Gene Symbol: LINC00607 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037195.1 | 4 | 546 | |||
| GenBank mRNA | AK091865.1 | - | 4 | 445 | ||
| AK124897.1 | - | 4 | 546 | |||