siRNA Details
| Gene Symbol: LINC00993 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104061.1 | 2 | 194 | |||
| GenBank mRNA | AI733546.1 | - | 2 | 209 | ||
| AI791937.1 | - | 2 | 84 | |||
| BX102924.1 | - | 2 | 194 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00993| Gene Symbol: LINC00993 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104061.1 | 2 | 194 | |||
| GenBank mRNA | AI733546.1 | - | 2 | 209 | ||
| AI791937.1 | - | 2 | 84 | |||
| BX102924.1 | - | 2 | 194 | |||