siRNA Details
| Gene Symbol: SPINK8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080525.1 | NP_001073994.1 | 3, 4 | 246 | ||
| XM_017007046.1 | XP_016862535.1 | 5, 6 | 836 | |||
| XM_017007047.1 | XP_016862536.1 | 5, 6 | 790 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SPINK8| Gene Symbol: SPINK8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080525.1 | NP_001073994.1 | 3, 4 | 246 | ||
| XM_017007046.1 | XP_016862535.1 | 5, 6 | 836 | |||
| XM_017007047.1 | XP_016862536.1 | 5, 6 | 790 | |||