siRNA Details
| Gene Symbol: FABP12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105281.1 | NP_001098751.1 | 2, 3 | 251 | ||
| XM_005251285.4 | XP_005251342.1 | 3, 4 | 920 | |||
| XM_006716465.3 | XP_006716528.1 | 3, 4 | 903 | |||
| XM_011517577.2 | XP_011515879.1 | 4, 5 | 1047 | |||
| GenBank mRNA | BC146973.1 | - | 2, 3 | 314 | 9056901 | |
| BC146981.1 | - | 2, 3 | 314 | 9056909 | ||
| EU733650.1 | - | 2, 3 | 262 | |||

FABP12
Genomic Map