siRNA Details
Gene Symbol: GYPE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017008139.1 | XP_016863628.1 | 1 | 440 | ||
XM_017008140.1 | XP_016863629.1 | 1 | 440 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: GYPE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017008139.1 | XP_016863628.1 | 1 | 440 | ||
XM_017008140.1 | XP_016863629.1 | 1 | 440 |