siRNA Details
Gene Symbol: HHIP-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037595.1 | 1, 2 | 309 | |||
GenBank mRNA | BC039450.1 | - | 1, 2 | 312 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: HHIP-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037595.1 | 1, 2 | 309 | |||
GenBank mRNA | BC039450.1 | - | 1, 2 | 312 |