siRNA Details
| Gene Symbol: LOC646626 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045484.1 | 2 | 832 | |||
| GenBank mRNA | BX104049.1 | - | 2 | 437 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC646626| Gene Symbol: LOC646626 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045484.1 | 2 | 832 | |||
| GenBank mRNA | BX104049.1 | - | 2 | 437 | ||